AMBH580647E6 Ambeed, Inc.

3-Bromo-1-ethyl-1H-indazole

  • MDL number MFCD20535074
  • Cas Number : 1352489-37-2
  • Molecular Weight : 225.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1352489-37-2.html

Safety Information

MSDS https://www.ambeed.com/products/1352489-37-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart