AMBH580647ED Ambeed, Inc.

3-Amino-6-bromopicolinamide

  • MDL number MFCD29059299
  • Cas Number : 1660117-37-2
  • Molecular Weight : 216.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1660117-37-2.html

Safety Information

MSDS https://www.ambeed.com/products/1660117-37-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart