AMBH580647F8 Ambeed, Inc.

7-Nitro-1H-benzo[d]imidazole

  • MDL number MFCD00220143
  • Cas Number : 10597-52-1
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/10597-52-1.html

Safety Information

MSDS https://www.ambeed.com/products/10597-52-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart