AMBH580647FE Ambeed, Inc.

(S)-3-(4-(Allyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

  • MDL number MFCD00273424
  • Cas Number : 127132-38-1
  • Molecular Weight : 321.3682

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/127132-38-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/127132-38-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart