AMBH580647FF Ambeed, Inc.

2-(Benzyloxy)-4-nitroaniline

  • MDL number MFCD00276800
  • Cas Number : 25945-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/25945-96-4.html

Safety Information

MSDS https://www.ambeed.com/products/25945-96-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart