AMBH58064808 Ambeed, Inc.

Brivanib

  • MDL number MFCD13194684
  • Cas Number : 649735-46-6
  • Molecular Weight : 370.3800

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/649735-46-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/649735-46-6.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart