AMBH5806480B Ambeed, Inc.

Sapropterin Dihydrochloride

  • MDL number MFCD00891665
  • Cas Number : 69056-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/69056-38-8.html

Safety Information

MSDS https://www.ambeed.com/products/69056-38-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart