AMBH5806480E Ambeed, Inc.

Methyl 4-((1H-1,2,4-triazol-3-yl)methyl)benzoate

  • MDL number MFCD26526981
  • Cas Number : 1785765-04-9
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-((1H-1,2,4-triazol-3-yl)methyl)benzoate
Inchi Code 1S/C11H11N3O2/c1-16-11(15)9-4-2-8(3-5-9)6-10-12-7-13-14-10/h2-5,7H,6H2,1H3,(H,12,13,14)
Inchi Key TYFFOCNYTODYNV-UHFFFAOYSA-N

Description

Synonym N/A
Purity 97%
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1785765-04-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1785765-04-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart