AMBH58064819 Ambeed, Inc.

Methyl 3-bromo-4-propoxybenzoate

  • MDL number MFCD06801019
  • Cas Number : 216965-51-4
  • Molecular Weight : 273.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/216965-51-4.html

Safety Information

MSDS https://www.ambeed.com/products/216965-51-4.html
Availability details for
Quantity
  • Availability
  • Add to Cart