AMBH5806481A Ambeed, Inc.

(3,3'-Difluoro-[2,2'-bithiophene]-5,5'-diyl)bis(trimethylstannane)

  • MDL number MFCD30543603
  • Cas Number : 1619967-09-7
  • Molecular Weight : 527.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1619967-09-7.html

Safety Information

MSDS https://www.ambeed.com/products/1619967-09-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart