AMBH58064820 Ambeed, Inc.

6-Chloro-2-iodopurine

  • MDL number MFCD09753499
  • Cas Number : 18552-90-4
  • Molecular Weight : 280.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-203°C
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 582.2°C at 760 mmHg
Product Link https://www.ambeed.com/products/18552-90-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/18552-90-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart