AMBH58064825 Ambeed, Inc.

A-1155463

  • MDL number MFCD29924714
  • Cas Number : 1235034-55-5
  • Molecular Weight : 669.7881

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1235034-55-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1235034-55-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart