AMBH58064829 Ambeed, Inc.

Sodium 4-chlorobenzenesulfinate

  • MDL number MFCD00035602
  • Cas Number : 14752-66-0
  • Molecular Weight : 198.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/14752-66-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14752-66-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart