AMBH58064834 Ambeed, Inc.

2-((2S,4R)-4-Hydroxypyrrolidin-2-yl)acetic acid hydrochloride

  • MDL number MFCD01862867
  • Cas Number : 336182-11-7
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/336182-11-7.html

Safety Information

MSDS https://www.ambeed.com/products/336182-11-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart