AMBH58064837 Ambeed, Inc.

1-BOC-3-trifluoromethyl-piperidin-4-one

  • MDL number MFCD08669340
  • Cas Number : 1159983-77-3
  • Molecular Weight : 267.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 130.7
Storage temp. Inert atmosphere,2-8°C
Shipping Temperature Normal
Country of Origin CN
Boiling point 292.5°C at 760 mmHg
Product Link https://www.ambeed.com/products/1159983-77-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1159983-77-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart