AMBH5806483D Ambeed, Inc.

2-(2-Phenyl-2H-tetrazol-5-yl)pyridine

  • MDL number N/A
  • Cas Number : 507270-06-6
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/507270-06-6.html

Safety Information

MSDS https://www.ambeed.com/products/507270-06-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart