AMBH58064842 Ambeed, Inc.

4-Chloro-2-(pyridin-2-yl)pyrimidine

  • MDL number MFCD11046846
  • Cas Number : 97603-38-8
  • Molecular Weight : 191.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/97603-38-8.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/97603-38-8.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart