AMBH5806484C Ambeed, Inc.

RET KInase inhibitor 1

  • MDL number MFCD30747847
  • Cas Number : 1627856-64-7
  • Molecular Weight : 467.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1627856-64-7.html

Safety Information

MSDS https://www.ambeed.com/products/1627856-64-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart