AMBH58064853 Ambeed, Inc.

MLN4924

  • MDL number MFCD17215201
  • Cas Number : 905579-51-3
  • Molecular Weight : 443.5193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/905579-51-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/905579-51-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart