AMBH58064854 Ambeed, Inc.

1A-116

  • MDL number MFCD31619254
  • Cas Number : 1430208-73-3
  • Molecular Weight : 307.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1430208-73-3.html

Safety Information

MSDS https://www.ambeed.com/products/1430208-73-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart