AMBH58064857 Ambeed, Inc.

Bis(trifluoromethane)sulfonimide lithium salt

  • MDL number MFCD00210017
  • Cas Number : 90076-65-6
  • Molecular Weight : 287.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/90076-65-6.html

Safety Information

signalword Danger
pcodes P273-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/90076-65-6.html
pictograms GHS05,GHS06,GHS08
hstatements H301+H311-H314-H318-H372-H412
Availability details for
Quantity
  • Availability
  • Add to Cart