AMBH58064858 Ambeed, Inc.

6-(3-(Piperidin-1-yl)propoxy)-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD31715275
  • Cas Number : 769920-49-2
  • Molecular Weight : 274.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(3-(piperidin-1-yl)propoxy)-1,2,3,4-tetrahydroquinoline
Inchi Code 1S/C17H26N2O/c1-2-10-19(11-3-1)12-5-13-20-16-7-8-17-15(14-16)6-4-9-18-17/h7-8,14,18H,1-6,9-13H2
Inchi Key CPRSNEKBYDXJTE-UHFFFAOYSA-N

Description

Synonym N/A
Purity 97%
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form Yellow to Brown Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/769920-49-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/769920-49-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart