AMBH58064863 Ambeed, Inc.

Ethyl 2-(5-bromo-2-methyl-1H-indol-3-yl)acetate

  • MDL number MFCD08272278
  • Cas Number : 72016-68-3
  • Molecular Weight : 296.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.ambeed.com/products/72016-68-3.html

Safety Information

MSDS https://www.ambeed.com/products/72016-68-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart