AMBH58064865 Ambeed, Inc.

5-Norbornene-2-carbonitrile

  • MDL number MFCD00167563
  • Cas Number : 95-11-4
  • Molecular Weight : 119.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 12-14
Iupac Name bicyclo[2.2.1]hept-5-ene-2-carbonitrile
Inchi Code 1S/C8H9N/c9-5-8-4-6-1-2-7(8)3-6/h1-2,6-8H,3-4H2
Inchi Key BMAXQTDMWYDIJX-UHFFFAOYSA-N

Description

Melting Point 12-14
Synonym N/A
Purity 98%
Flash Point 66
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/95-11-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/95-11-4.html
pictograms GHS07
hstatements H227-H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart