AMBH58064869 Ambeed, Inc.

7-Bromoimidazo[1,2-a]pyrimidine

  • MDL number MFCD16877232
  • Cas Number : 1251033-57-4
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromoimidazo[1,2-a]pyrimidine
Inchi Code 1S/C6H4BrN3/c7-5-1-3-10-4-2-8-6(10)9-5/h1-4H
Inchi Key QNMPFHNDEOTXNR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Product Link https://www.ambeed.com/products/1251033-57-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1251033-57-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart