AMBH5806486A Ambeed, Inc.

Varenicline Tartrate

  • MDL number MFCD08460603
  • Cas Number : 375815-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/375815-87-5.html

Safety Information

MSDS https://www.ambeed.com/products/375815-87-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart