AMBH5806486D Ambeed, Inc.

2-(Piperidin-4-ylidene)acetic acid hydrochloride

  • MDL number MFCD11506216
  • Cas Number : 84839-57-6
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/84839-57-6.html

Safety Information

MSDS https://www.ambeed.com/products/84839-57-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart