AMBH58064876 Ambeed, Inc.

Ethyl 4,4,4-trifluorocrotonate

  • MDL number MFCD00009903
  • Cas Number : 25597-16-4
  • Molecular Weight : 168.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/25597-16-4.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/25597-16-4.html
pictograms GHS02,GHS07
hstatements H226-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart