AMBH5806487C Ambeed, Inc.

1-(Chloromethyl)-2,3-difluorobenzene

  • MDL number MFCD03095320
  • Cas Number : 446-57-1
  • Molecular Weight : 162.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 176.2�t 760 mmHg
Iupac Name 1-(chloromethyl)-2,3-difluorobenzene
Inchi Code 1S/C7H5ClF2/c8-4-5-2-1-3-6(9)7(5)10/h1-3H,4H2
Inchi Key OVUKQQRPTLPXTD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Inert atmosphere,2-8
Physical Form Solid or liquid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 176.2�t 760 mmHg
Product Link https://www.ambeed.com/products/446-57-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/446-57-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart