AMBH58064885 Ambeed, Inc.

(R)-Methyl pyrrolidine-2-carboxylate

  • MDL number MFCD06656665
  • Cas Number : 43041-12-9
  • Molecular Weight : 129.1570

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form Solid or liquid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/43041-12-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/43041-12-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart