AMBH58064888 Ambeed, Inc.

tert-Butyl 1-hydroxy-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate

  • MDL number MFCD11041105
  • Cas Number : 1334177-84-2
  • Molecular Weight : 498.6048

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1334177-84-2.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1334177-84-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart