AMBH58064889 Ambeed, Inc.

Fmoc-Phe(3-CF3)-OH

  • MDL number MFCD00672555
  • Cas Number : 205526-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/205526-27-8.html

Safety Information

MSDS https://www.ambeed.com/products/205526-27-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart