AMBH58064894 Ambeed, Inc.

Zanamivir hydrate

  • MDL number MFCD03791324
  • Cas Number : 1260601-68-0
  • Molecular Weight : 350.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1260601-68-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1260601-68-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart