AMBH58064897 Ambeed, Inc.

5-(4-Ethylpiperazin-1-yl)pyridin-2-amine

  • MDL number MFCD10009575
  • Cas Number : 1018505-59-3
  • Molecular Weight : 206.2874

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1018505-59-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1018505-59-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart