AMBH58064899 Ambeed, Inc.

2-(2-(Diethylamino)ethoxy)ethanol

  • MDL number MFCD00020604
  • Cas Number : 140-82-9
  • Molecular Weight : 161.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 51.3
Storage temp. Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 161.2°C at 760 mmHg
Product Link https://www.ambeed.com/products/140-82-9.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
hcodes 8
MSDS https://www.ambeed.com/products/140-82-9.html
pictograms GHS05,GHS07
hstatements H312-H314
Availability details for
Quantity
  • Availability
  • Add to Cart