AMBH580648A9 Ambeed, Inc.

SHP099

  • MDL number N/A
  • Cas Number : 1801747-42-1
  • Molecular Weight : 352.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1801747-42-1.html

Safety Information

MSDS https://www.ambeed.com/products/1801747-42-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart