AMBH580648AC Ambeed, Inc.

3-((2S,3S)-3-Amino-1-(4-chlorophenyl)butan-2-yl)benzonitrile hydrochloride

  • MDL number MFCD11042327
  • Cas Number : 610791-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Room Temp
Physical Form solid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/610791-48-5.html

Safety Information

MSDS https://www.ambeed.com/products/610791-48-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart