AMBH580648B3 Ambeed, Inc.

Ethyl (R)-2-hydroxy-4-phenylbutyrate

  • MDL number MFCD00077794
  • Cas Number : 90315-82-5
  • Molecular Weight : 208.2536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/90315-82-5.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/90315-82-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart