AMBH580648B5 Ambeed, Inc.

(S)-1-(6-Bromopyridin-3-yl)ethanamine

  • MDL number MFCD09256775
  • Cas Number : 1213594-37-6
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1213594-37-6.html

Safety Information

MSDS https://www.ambeed.com/products/1213594-37-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart