AMBH580648BF Ambeed, Inc.

Methyl 5-methoxy-2,3-dihydro-1H-indene-2-carboxylate

  • MDL number MFCD21363132
  • Cas Number : 65844-46-4
  • Molecular Weight : 206.2378

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/65844-46-4.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/65844-46-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart