AMBH580648C0 Ambeed, Inc.

2,2'-[PRopane-2,2-diylbis(thio)]diacetic acid

  • MDL number MFCD01830380
  • Cas Number : 4265-58-1
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2'-(propane-2,2-diylbis(sulfanediyl))diacetic acid
Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/4265-58-1.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/4265-58-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart