AMBH580648C1 Ambeed, Inc.

AT406

  • MDL number MFCD22124467
  • Cas Number : 1071992-99-8
  • Molecular Weight : 561.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1071992-99-8.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1071992-99-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart