AMBH580648C4 Ambeed, Inc.

1-(Bromomethyl)-3-fluoro-5-(trifluoromethyl)benzene

  • MDL number MFCD00061175
  • Cas Number : 239087-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Room Temp
Physical Form liquid
Shipping Temperature Room Temp
Country of Origin China
Product Link https://www.ambeed.com/products/239087-09-3.html

Safety Information

MSDS https://www.ambeed.com/products/239087-09-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart