AMBH580648C8 Ambeed, Inc.

3-Formylpicolinonitrile

  • MDL number MFCD07367918
  • Cas Number : 131747-66-5
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/131747-66-5.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P311
MSDS https://www.ambeed.com/products/131747-66-5.html
pictograms GHS06
hstatements H302+H312-H315-H319-H331-H335
Availability details for
Quantity
  • Availability
  • Add to Cart