AMBH580648C9 Ambeed, Inc.

Ethyl dimethylcarbamate

  • MDL number MFCD01738220
  • Cas Number : 687-48-9
  • Molecular Weight : 117.1463

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/687-48-9.html

Safety Information

signalword Danger
pcodes P210-P261-P305+P351+P338
MSDS https://www.ambeed.com/products/687-48-9.html
pictograms GHS02,GHS07
hstatements H226-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart