AMBH580648CA Ambeed, Inc.

Cerium hydrate

  • MDL number MFCD00010927
  • Cas Number : 12014-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/12014-56-1.html

Safety Information

MSDS https://www.ambeed.com/products/12014-56-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart