AMBH580648D2 Ambeed, Inc.

Nicotinic acid riboside

  • MDL number MFCD30730071
  • Cas Number : 17720-18-2
  • Molecular Weight : 255.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/17720-18-2.html

Safety Information

MSDS https://www.ambeed.com/products/17720-18-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart