AMBH580648D3 Ambeed, Inc.

Cinnamyl chloride

  • MDL number MFCD00000986
  • Cas Number : 2687-12-9
  • Molecular Weight : 152.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2687-12-9.html

Safety Information

signalword Danger
pcodes P260-P280-P284-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/2687-12-9.html
pictograms GHS05,GHS06,GHS08
hstatements H302-H314-H330-H334
Availability details for
Quantity
  • Availability
  • Add to Cart