AMBH580648D5 Ambeed, Inc.

(S)-2-amino-6-(3-(3-(3-methyl-3H-diazirin-3-yl)propyl)ureido)hexanoic acid

  • MDL number MFCD28900721
  • Cas Number : 1337883-32-5
  • Molecular Weight : 285.3427

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1337883-32-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1337883-32-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart