AMBH580648D9 Ambeed, Inc.

(2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-mercapto-9H-purin-9-yl)tetrahydrofuran-3,4-diol

  • MDL number MFCD00005743
  • Cas Number : 574-25-4
  • Molecular Weight : 284.2917

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/574-25-4.html

Safety Information

signalword Warning
pcodes P264-P501-P301+P312+P330-P270
MSDS https://www.ambeed.com/products/574-25-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart